Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRCAA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103112
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103112 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103112 Supplier Invitrogen™ Supplier No. PA5103112
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BRCAA1.

The exact function of C18orf25 remains unknown. There are two named isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCAA1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ARID4B
Gene Accession No. A2CG63, Q4LE39, Q9JKB5
Gene Alias 180 kDa Sin3-associated polypeptide; ARID domain-containing protein 4B; ARID4B; AT rich interactive domain 4B (Rbp1 like); AT rich interactive domain 4B (RBP1-like); AT-rich interaction domain 4B; AT-rich interactive domain-containing protein 4B; Bcaa; BRCAA1; breast cancer-associated antigen; breast cancer-associated antigen 1; breast cancer-associated antigen BRCAA1; breast carcinoma-associated antigen; histone deacetylase complex subunit SAP180; Rb-binding protein homolog; RBBP1L1; RBP1L1; retinoblastoma binding protein 1-like 1; retinoblastoma-binding protein 1-like 1; retinoblastoma-binding protein 1-related protein; Sap180; SIN3A-associated protein 180; Sin3-associated polypeptide p180
Gene Symbols ARID4B
Host Species Rabbit
Immunogen A synthesized peptide derived from human ARID4B(Accession Q4LE39), corresponding to amino acid residues S1278-R1312.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51742, 84481, 94246
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.