Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRCAA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139781 Supplier Invitrogen™ Supplier No. PA5139781
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: NCKLRRLSKP PFQTNPSPEM VSKLDLTDAK NSDTAHIKSI EITSILNGLQ ASESSAEDSE QEDERGAQDM Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 91%.

The exact function of C18orf25 remains unknown. There are two named isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRCAA1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARID4B
Gene Accession No. Q4LE39
Gene Alias 180 kDa Sin3-associated polypeptide; ARID domain-containing protein 4B; ARID4B; AT rich interactive domain 4B (Rbp1 like); AT rich interactive domain 4B (RBP1-like); AT-rich interaction domain 4B; AT-rich interactive domain-containing protein 4B; Bcaa; BRCAA1; breast cancer-associated antigen; breast cancer-associated antigen 1; breast cancer-associated antigen BRCAA1; breast carcinoma-associated antigen; histone deacetylase complex subunit SAP180; Rb-binding protein homolog; RBBP1L1; RBP1L1; retinoblastoma binding protein 1-like 1; retinoblastoma-binding protein 1-like 1; retinoblastoma-binding protein 1-related protein; Sap180; SIN3A-associated protein 180; Sin3-associated polypeptide p180
Gene Symbols ARID4B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARID4B. Recombinant protein control fragment (Product #RP-108765).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51742
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.