Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRCAA1 Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502277982
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7982 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7982 Supplier Leinco Technologies Inc Supplier No. 34022100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BRCAA1 Polyclonal antibody specifically detects BRCAA1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen BRCAA1
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ARID4B
Gene Accession No. A2CG63, Q4LE39, Q9JKB5
Gene Alias 180 kDa Sin3-associated polypeptide; ARID domain-containing protein 4B; ARID4B; AT rich interactive domain 4B (Rbp1 like); AT rich interactive domain 4B (RBP1-like); AT-rich interaction domain 4B; AT-rich interactive domain-containing protein 4B; Bcaa; BRCAA1; breast cancer-associated antigen; breast cancer-associated antigen 1; breast cancer-associated antigen BRCAA1; breast carcinoma-associated antigen; histone deacetylase complex subunit SAP180; Rb-binding protein homolog; RBBP1L1; RBP1L1; retinoblastoma binding protein 1-like 1; retinoblastoma-binding protein 1-like 1; retinoblastoma-binding protein 1-related protein; Sap180; SIN3A-associated protein 180; Sin3-associated polypeptide p180
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 1046-1055 (SSKKQKRSHK) of human BRCAA1 conjugated to KLH.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51742, 84481, 94246
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.