Learn More
Invitrogen™ BRCC3 Polyclonal Antibody

Description
A suggested positive control is MCF7 cell lysate. PA5-20426 can be used with blocking peptide PEP-0543.
Specifications
Specifications
| Antigen | BRCC3 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | BRCC3 |
| Gene Accession No. | B2RYM5, P46736, P46737 |
| Gene Alias | 6.1A protein; BRCA1/BRCA2-containing complex subunit 3; BRCA1/BRCA2-containing complex subunit 36; BRCA1/BRCA2-containing complex, subunit 3; BRCA1-A complex subunit BRCC36; Brcc3; BRCC36; BRISC complex subunit BRCC36; C6.1A; CXorf53; lys-63-specific deubiquitinase BRCC36; RP11-143H17.2 |
| Gene Symbols | BRCC3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.