Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BRD 0539
GREENER_CHOICE

Catalog No. 715310
Change view
Click to view available options
Quantity:
2 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-531-0 10 mg
71-532 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 71-531-0 Supplier Tocris Bioscience™ Supplier No. 7153/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cell permeable and reversible Cas9 inhibitor

BRD 0539 is a cell-permeable and reversible SpCas9 (Streptococcus pyogenes Cas9) inhibitor. BRD 0539 shows dose-dependent SpCas9 inhibition in an in vitro DNA cleavage assay (apparent IC50 = 22 μM) and in an eGFP disruption assay (apparent EC50 = 11 μM). Allows dose and temporal control of SpCas9-based systems.

Specifications

Chemical Name or Material (3 aR,4 R,9 bR)-8-(2-Fluorophenyl)-2,3,3 A,4,5,9 b-hexahydro-1-[(4-methylphenyl)sulfonyl]-1 H-pyrrolo[3,2-c]quinoline-4-methanol
CAS 1403838-79-8
Quantity 10 mg
Target CRISPR Reagents
Molecular Formula C25H25FN2O3S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.