Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BRD 4770

Catalog No. p-8456002
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-821-0 10 mg
62-825-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-821-0 Supplier Tocris Bioscience™ Supplier No. 6282/10
This item is not returnable. View return policy

G9a inhibitor and S-adenosyl methionine mimetic; cell permeable

BRD 4770 is a G9a inhibitor and S-adenosyl methionine (SAM) mimetic. Inhibits methylation of H3K9 (EC50 ~ 5 μM) and induces senescence in PANC-1 cells. Induces ATM activation without DNA damage. Cell permeable.

Specifications

Chemical Name or Material Methyl-2-(benzoylamino)-1-(3-phenylpropyl)-1H-benzimidazole-5-carboxylate
CAS 1374601-40-7
Assay Percent Range >98%
Quantity 10 mg
Molecular Formula C25H23N3O3
Formula Weight Observed MW: 413.47
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.