Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BRD 73954
GREENER_CHOICESDP

Catalog No. 628450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
62-845-0 50 mg
62-841-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 62-845-0 Supplier Tocris Bioscience™ Supplier No. 6284/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dual histone deacetylase (HDAC) 6/8 inhibitor

BRD 73954 is a dual histone deacetylase (HDAC) 6/8 inhibitor. IC50 values are 0.036 μM, 0.12 μM, 9 μM, >33 μM, for HDAC6, HDAC8, HDAC2 and HDAC4, respectively.

Specifications

Chemical Name or Material N1-Hydroxy-N3-(2-phenylethyl)-1,3-benzenedicarboxamide
CAS 1440209-96-0
Assay Percent Range >98%
Quantity 50 mg
Molecular Formula C16H16N2O3
Formula Weight Observed MW: 284.31
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.