Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA581916
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA581916 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA581916 Supplier Invitrogen™ Supplier No. PA581916
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IAAEVGQSSM WISTDAAASV LEPLKVVWAK CSGYPSYPAL IIDPKMPRVP GHHNGVTIPA PPLDVLKIGE HMQTKSDEKL FLVLFFDNKR SWQWLPKSKM VPLGIDETID KLKMMEGRNS SIRKAVRIAF DRAMNHLSRV HGEPTSD Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 97%.

This gene encodes a protein of unknown function. The protein contains a bromodomain, a sequence motif often found in transcriptional coactivators, and localizes to the nucleus in testis and several other cell types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRD1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BRD1
Gene Accession No. O95696
Gene Alias 1110059H06Rik; AI316859; BR140-like protein; BRD1; BRL; Bromodomain and PHD finger-containing protein 2; bromodomain containing 1; bromodomain-containing protein 1; BRPF1; BRPF2; DKFZp686F0325; mKIAA4191; RP3-522J7.4
Gene Symbols BRD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BRD1. Recombinant protein control fragment (Product #RP-91380).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23774
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.