Learn More
BRD1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302366A

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein of unknown function. The protein contains a bromodomain, a sequence motif often found in transcriptional coactivators, and localizes to the nucleus in testis and several other cell types.Specifications
BRD1 | |
Polyclonal | |
Unconjugated | |
BRD1 | |
RP3-522J7.4, BRL, BRPF1, BRPF2, DKFZp686F0325, BR140-like protein; bromodomain and PHD finger-containing protein 2; bromodomain-containing protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23774 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
O95696 | |
BRD1 | |
Between 750 and 800 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.