Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988554
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8554 Supplier Bioss Supplier No. BS8426R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein of unknown function. The protein contains a bromodomain, a sequence motif often found in transcriptional coactivators, and localizes to the nucleus in testis and several other cell types.

Specifications

Antigen BRD1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BRD1
Gene Accession No. O95696
Gene Alias 1110059H06Rik; AI316859; BR140-like protein; BRD1; BRL; Bromodomain and PHD finger-containing protein 2; bromodomain containing 1; Bromodomain-containing protein 1; BRPF1; BRPF2; DKFZp686F0325; mKIAA4191; RP3-522J7.4
Gene Symbols BRD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BRD1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23774
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.