Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRD2 Antibody (7C1B10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | BRD2 |
---|---|
Clone | 7C1B10 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61716
![]() |
Novus Biologicals
NBP261716 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
BRD2 Monoclonal specifically detects BRD2 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
BRD2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human, Rat | |
6046 | |
Purified recombinant fragment of human BRD2 (AA: 227-364) expressed in E. Coli. | |
Primary | |
45.5 kDa |
7C1B10 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
Monoclonal | |
Purified | |
Cellular Markers, DNA Repair, Protein Kinase | |
bromodomain containing 2, bromodomain-containing 2, bromodomain-containing protein 2, D6S113E, female sterile homeotic-related gene 1, FSRG1FSH, KIAA9001FLJ31942, NAT, O27.1.1, Really interesting new gene 3 protein, RING3DKFZp686N0336, RNF3 | |
BRD2 | |
IgG2b | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title