Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRD2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130474
Description
BRD2 Polyclonal specifically detects BRD2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
BRD2 | |
Polyclonal | |
Unconjugated | |
Tris-buffered Saline with 0.09% Sodium Azide | |
BRD2 | |
The immunogen for this product maps to a region between residue 1 and 50 of human bromodomain containing 2 using the numbering given in entry NP_005095.1 (GeneID 6046). | |
0.1 mL | |
Cellular Markers, DNA Repair, Protein Kinase | |
6046 | |
Store at 4C. Do not freeze. |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 2-5 ug/mg lysate | |
bromodomain containing 2, bromodomain-containing 2, bromodomain-containing protein 2, D6S113E, female sterile homeotic-related gene 1, FSRG1FSH, KIAA9001FLJ31942, NAT, O27.1.1, Really interesting new gene 3 protein, RING3DKFZp686N0336, RNF3 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction