Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRD2 Monoclonal Antibody (1H6B12)
GREENER_CHOICE

Catalog No. PIMA531655
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531655 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531655 Supplier Invitrogen™ Supplier No. MA531655
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31655 has been tested in indirect ELISA.

This gene encodes a transcriptional regulator that belongs to the BET (bromodomains and extra terminal domain) family of proteins. This protein associates with transcription complexes and with acetylated chromatin during mitosis, and it selectively binds to the acetylated lysine-12 residue of histone H4 via its two bromodomains. The gene maps to the major histocompatibility complex (MHC) class II region on chromosome 6p21. 3, but sequence comparison suggests that the protein is not involved in the immune response. This gene has been implicated in juvenile myoclonic epilepsy, a common form of epilepsy that becomes apparent in adolescence. Multiple alternatively spliced variants have been described for this gene, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRD2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1H6B12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene BRD2
Gene Accession No. P25440
Gene Alias AW228947; Brd2; bromodomain containing 2; bromodomain-containing 2; bromodomain-containing protein 2; D17H6S113E; D6S113E; DADB-17J1.3; DKFZp686N0336; female sterile homeotic-related gene 1; Female sterile homeotic-related protein 1; FLJ31942; Frg-1; FSH; Fsrg1; Fsrg-1; Kiaa4005; KIAA9001; mKIAA4005; Nat; O27.1.1; Protein RING3; Really interesting new gene 3 protein; RING3; RING3 protein; Rnf3
Gene Symbols BRD2
Host Species Mouse
Immunogen Purified recombinant fragment of human BRD2 (AA: 227-364) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6046
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.