Learn More
BRD2 Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | BRD2 |
| Applications | ChIP sequencing (ChIP-seq) |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | BRD2 |
| Gene Accession No. | P25440 |
| Gene Alias | AW228947; BRD2; bromodomain containing 2; bromodomain-containing 2; Bromodomain-containing protein 2; D17H6S113E; D6S113E; DADB-17J1.3; DKFZp686N0336; female sterile homeotic-related gene 1; Female sterile homeotic-related protein 1; FLJ31942; Frg-1; FSH; Fsrg1; Fsrg-1; Kiaa4005; KIAA9001; mKIAA4005; NAT; O27.1.1; Protein RING3; Really interesting new gene 3 protein; Ring3; RING3 protein; Rnf3 |
| Gene Symbols | BRD2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.