Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRD2 Recombinant Rabbit Monoclonal Antibody (BL-167-2A2)

Catalog No. PIMA544212
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544212 20 μL
PIMA544211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544212 Supplier Invitrogen™ Supplier No. MA544212
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

BRD2 Recombinant Monoclonal Antibody for Western Blot, IP

This gene encodes a transcriptional regulator that belongs to the BET (bromodomains and extra terminal domain) family of proteins. This protein associates with transcription complexes and with acetylated chromatin during mitosis, and it selectively binds to the acetylated lysine-12 residue of histone H4 via its two bromodomains. The gene maps to the major histocompatibility complex (MHC) class II region on chromosome 6p21. 3, but sequence comparison suggests that the protein is not involved in the immune response. This gene has been implicated in juvenile myoclonic epilepsy, a common form of epilepsy that becomes apparent in adolescence. Multiple alternatively spliced variants have been described for this gene, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRD2
Applications Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone BL-167-2A2
Concentration 40 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
Gene BRD2
Gene Accession No. P25440, Q7JJ13
Gene Alias AW228947; Brd2; bromodomain containing 2; bromodomain-containing 2; bromodomain-containing protein 2; D17H6S113E; D6S113E; DADB-17J1.3; DKFZp686N0336; female sterile homeotic-related gene 1; Female sterile homeotic-related protein 1; FLJ31942; Frg-1; FSH; Fsrg1; Fsrg-1; Kiaa4005; KIAA9001; mKIAA4005; Nat; O27.1.1; Protein RING3; Really interesting new gene 3 protein; RING3; RING3 protein; Rnf3
Gene Symbols BRD2
Host Species Rabbit
Immunogen Synthetic peptide representing a region between residues 675 and 725 of human bromodomain containing 2.
Purification Method Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14312, 6046
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.