Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD2 Recombinant Rabbit Monoclonal Antibody (BL-167-2A2), Bethyl Laboratories

Catalog No. 501571269
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
20 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-1269 100 μL
50-157-1270 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-1269 Supplier Bethyl Laboratories, Inc Supplier No. A700008
Add to Cart
Add to Cart

Rabbit Recombit Monoclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

BRD2 is a mitogen-activated kinase which localizes to the nucleus. The gene maps to the major histocompatability complex (MHC) class II region on chromosome 6p21.3 but sequence comparison suggests that the protein is not involved in the immune response. Homology to the Drosophila gene female sterile homeotic suggests that this human protein may be part of a signal transduction pathway involved in growth control.

Specifications

Antigen BRD2
Applications Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone BL-167-2A2
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
Gene BRD2
Gene Accession No. P25440
Gene Alias DADB-17J1.3, D6S113E, DKFZp686N0336, FLJ31942, FSH, FSRG1, KIAA9001, NAT, RING3, RNF3
Gene Symbols BRD2
Host Species Rabbit
Immunogen Between 675 and 725
Purification Method Ion-exchange chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6046
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.