Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD3 Polyclonal Antibody, Active Motif™
SDP

Rabbit Polyclonal Antibody

$159.90 - $651.90

Specifications

Antigen BRD3
Concentration Lot-Specific
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3443
SDP
Encompass
View Documents
Active Motif
61490
10 μL
Each for $159.90
Only null left
Add to Cart
 
50-199-3442
SDP
Encompass
View Documents
Active Motif
61489
100 μL
Each for $651.90
Only null left
Add to Cart
 
Description

Description

This gene was identified based on its homology to the gene encoding the RING3 protein, a serine/threonine kinase. The gene localizes to 9q34, a region which contains several major histocompatibility complex (MHC) genes. The function of the encoded protein is not known.
Specifications

Specifications

BRD3
ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Unconjugated
Rabbit
Human, Mouse
Q15059, Q8K2F0
67382, 8019
This antibody was raised against a peptide within the C-terminal region of mouse BRD3.
Primary
-20°C, Avoid Freeze/Thaw Cycles
BRD3
Lot-Specific
Polyclonal
Liquid
RUO
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
2410084F24Rik; AW060456; BOS_12102; Brd3; bromodomain containing 3; bromodomain containing protein 3; bromodomain-containing 3; Bromodomain-containing FSH-like protein FSRG2; Bromodomain-containing protein 3; bromodomain-containing protein 3 short isoform; Fsrg2; KIAA0043; ORFX; RING3L; RING3-like protein; RINGL3
BRD3
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.