Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRD3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982449
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2449 Supplier Bioss Supplier No. BS12889R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene was identified based on its homology to the gene encoding the RING3 protein, a serine/threonine kinase. The gene localizes to 9q34, a region which contains several major histocompatibility complex (MHC) genes. The function of the encoded protein is not known.

Specifications

Antigen BRD3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene BRD3
Gene Accession No. Q15059
Gene Alias 2410084F24Rik; AW060456; BOS_12102; Brd3; bromodomain containing 3; bromodomain containing protein 3; bromodomain-containing 3; Bromodomain-containing FSH-like protein FSRG2; Bromodomain-containing protein 3; bromodomain-containing protein 3 short isoform; Fsrg2; KIAA0043; ORFX; RING3L; RING3-like protein; RINGL3
Gene Symbols BRD3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human BRD3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8019
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.