Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRD3 Recombinant Rabbit Monoclonal Antibody (BLR069G)
GREENER_CHOICE

Catalog No. PIMA544329
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544329 100 μL
PIMA544330 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544329 Supplier Invitrogen™ Supplier No. MA544329
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.

This gene was identified based on its homology to the gene encoding the RING3 protein, a serine/threonine kinase. The gene localizes to 9q34, a region which contains several major histocompatibility complex (MHC) genes. The function of the encoded protein is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRD3
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BLR069G
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
Gene BRD3
Gene Accession No. Q15059
Gene Alias 2410084F24Rik; AW060456; BRD3; bromodomain containing 3; bromodomain containing 3; bromodomain-containing protein 3; bromodomain containing protein 3; bromodomain-containing 3; bromodomain-containing FSH-like protein FSRG2; Bromodomain-containing protein 3; bromodomain-containing protein 3 short isoform; Fsrg2; KIAA0043; ORFX; RING3L; RING3-like protein; RINGL3
Gene Symbols BRD3
Host Species Rabbit
Immunogen Peptide representing a region between residue 676 and 726 of human Bromodomain-containing protein 3 (NP_031397.1).
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8019
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.