Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRD4 Antibody (BL-149-2H5), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP276393
Description
BRD4 Monoclonal specifically detects BRD4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation Sequencing.Specifications
BRD4 | |
Monoclonal | |
Unconjugated | |
Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA with 0.09% Sodium Azide | |
BRD4 | |
Immunogen sequence between amino acids 1312 and 1362 | |
100 μL | |
23476.0 | |
Store at 2-8°C/ 1 year from date of receipt | |
IgG |
ChIP Assay, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
BL-149-2H5 | |
Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:25 - 1:250, Immunocytochemistry/Immunofluorescence 1:25-1:250, Immunoprecipitation 5 - 20 μL/0.5 mg lysate, Immunohistochemistry-Paraffin, Chromatin Immunoprecipitation Sequencing | |
bromodomain containing 4, bromodomain-containing 4, CAPchromosome-associated protein, HUNK1bromodomain-containing protein 4, HUNKI, MCAP, Protein HUNK1 | |
Rabbit | |
Antigen Affinity-purified | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction