Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRD4 Antibody (RAB-C131), Alexa Fluor™ 532 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320125AF532
Description
BRD4 Monoclonal antibody specifically detects BRD4 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
BRD4 | |
Monoclonal | |
Alexa Fluor 532 | |
bromodomain containing 4, bromodomain-containing 4, CAPchromosome-associated protein, HUNK1bromodomain-containing protein 4, HUNKI, MCAP, Protein HUNK1 | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing BRD4 protein under non-denaturing conditions. | |
0.1 mL | |
Cancer, Signal Transduction | |
23476 | |
Store at 4&drg;C in the dark. | |
IgG κ |
Flow Cytometry, ELISA, Immunofluorescence | |
RAB-C131 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction