Learn More
Invitrogen™ BRD4 Chimeric Recombinant Rabbit Monoclonal Antibody (RAB-C131)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA548328
Description
Specificity: This antibody recognizes BRD4 (Bromodomain-containing protein 4). It binds to a folded domain, amino acids 333-460. BRD4 is a chromatin reader protein that recognizes and binds acetylated histones and plays a key role in transmission of epigenetic memory.
BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.
Specifications
BRD4 Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
O60885 | |
BRD4 | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing BRD4 protein under non-denaturing conditions. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG κ |
ChIP Assay, ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
RAB-C131 | |
Unconjugated | |
BRD4 | |
Brd4; BRD4-NUT FUSION; Brd5; bromodomain containing 4; bromodomain-containing 4; bromodomain-containing 5; bromodomain-containing protein 4; bromodomain-containing protein 4; LOW QUALITY PROTEIN: bromodomain-containing protein 4; CAP; chromosome-associated protein; fsh-like protein; HUNK1; HUNKI; Mcap; mitotic chromosome-associated protein; Protein HUNK1; WI-11513 | |
Rabbit | |
Protein A | |
RUO | |
23476 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.