Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRD4 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP275089
Description
BRD4 Polyclonal specifically detects BRD4 in Mouse samples. It is validated for Western Blot, Peptide ELISA.Specifications
BRD4 | |
Polyclonal | |
Unconjugated | |
TBS (pH 7.3), 0.5% BSA with 0.02% Sodium Azide | |
BRD4 | |
Peptide with sequence C-PETSNPNKPKRQTNQ, from the internal region (near N Terminus) of the protein sequence according to NP_490597.1; NP_055114.1. | |
Antigen Affinity-purified | |
Primary | |
Mouse | |
Purified |
Western Blot, ELISA | |
0.5 mg/ml | |
Western Blot 0.5-2 ug/ml, Peptide ELISA 1:8000 | |
bromodomain containing 4, bromodomain-containing 4, CAPchromosome-associated protein, HUNK1bromodomain-containing protein 4, HUNKI, MCAP, Protein HUNK1 | |
Goat | |
155.8 kDa | |
100 μg | |
23476.0 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction