Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRD4 Rabbit anti-Human, Active Motif™

Rabbit Recombinant Monoclonal Antibody
$108.45 - $482.00
Specifications
Antigen | BRD4 |
---|---|
Concentration | 1 mg/mL |
Applications | ChIP sequencing (ChIP-seq) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3868
![]() ![]() |
Active Motif
91302 |
10 μg |
Each for $108.45
|
|
|||||
50-199-3867
![]() ![]() |
Active Motif
91301 |
100 μg |
Each for $482.00
|
|
|||||
Description
BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.Specifications
BRD4 | |
ChIP sequencing (ChIP-seq) | |
Unconjugated | |
Rabbit | |
Human | |
O60885 | |
23476 | |
This antibody was raised against a recombinant protein comprising amino acids 149-284 of human BRD4. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
BRD4 |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 | |
BOS_7204; Brd4; BRD4-NUT FUSION; Brd5; bromodomain containing 4; bromodomain-containing 4; bromodomain-containing 5; Bromodomain-containing protein 4; CAP; chromosome-associated protein; fsh-like protein; HUNK1; HUNKI; Mcap; Mitotic chromosome-associated protein; Protein HUNK1; WI-11513 | |
BRD4 | |
IgG | |
Protein A | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title