Learn More
BRD4 Rabbit anti-Human, Polyclonal , Abnova™
BRD4 polyclonal antibody raised against recombinant human BRD4.
Supplier: Abnova Corporation PAB29479
Description
The protein encoded by this gene is homologous to the murine protein MCAP, which associates with chromosomes during mitosis, and to the human RING3 protein, a serine/threonine kinase. Each of these proteins contains two bromodomains, a conserved sequence motif which may be involved in chromatin targeting. This gene has been implicated as the chromosome 19 target of translocation t(15;19)(q13;p13.1), which defines an upper respiratory tract carcinoma in young people. Two alternatively spliced transcript variants have been described. [provided by RefSeq]
Sequence: PPFIATQVPVLEPQLPGSVFDPIGHFTQPILHLPQPELPPHLPQPPEHSTPPHLNQHAVVSPPALHNALPQQPSRPSNSpecifications
BRD4 | |
Polyclonal | |
BRD4 polyclonal antibody raised against recombinant human BRD4. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
BRD4 | |
Recombinant protein corresponding to amino acids of human BRD4. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunofluorescence (1-4 ug/mL) Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
CAP/HUNK1/HUNKI/MCAP | |
Rabbit | |
Antigen affinity purification | |
RUO | |
23476 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.