Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRD4 Recombinant Rabbit Monoclonal Antibody (BL-151-6F11)
GREENER_CHOICE

Catalog No. PIMA544205
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544205 100 μL
PIMA544206 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544205 Supplier Invitrogen™ Supplier No. MA544205
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

BRD4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, IP

BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRD4
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BL-151-6F11
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
Gene BRD4
Gene Accession No. O60885, Q9ESU6
Gene Alias Brd4; BRD4-NUT FUSION; Brd5; bromodomain containing 4; bromodomain-containing 4; bromodomain-containing 5; bromodomain-containing protein 4; bromodomain-containing protein 4; LOW QUALITY PROTEIN: bromodomain-containing protein 4; CAP; chromosome-associated protein; fsh-like protein; HUNK1; HUNKI; Mcap; mitotic chromosome-associated protein; Protein HUNK1; WI-11513
Gene Symbols BRD4
Host Species Rabbit
Immunogen Synthetic peptide representing a region between residues 1312 and 1362 of human bromodomain-containing protein 4.
Purification Method purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23476, 57261
Target Species Human, Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.