Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRD4-S Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116913
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116913 Supplier Invitrogen™ Supplier No. PA5116913
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, HeLa-BRD4-FL, HeLa-BRD4-S(a), HeLa-BRD4-S(b) Immunogen sequence: ESEEEDKCKP MSYEEKRQLS LDINKLPGEK LGRVVHIIQS REPSLKNSNP DEIEIDFETL KPSTLRELER YVTSCLRKKR KPQAEKVDVI AGSSKMKGFS SSESESSSES SSSDSEDSET GPA.

The protein encoded by this gene is homologous to the murine protein MCAP, which associates with chromosomes during mitosis, and to the human RING3 protein, a serine/threonine kinase. Each of these proteins contains two bromodomains, a conserved sequence motif which may be involved in chromatin targeting. This gene has been implicated as the chromosome 19 target of translocation t(15;19)(q13;p13.1), which defines an upper respiratory tract carcinoma in young people. Two alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRD4-S
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene BRD4
Gene Accession No. O60885
Gene Alias BRD4; bromodomain containing 4; bromodomain-containing 4; bromodomain-containing protein 4; CAP; chromosome-associated protein; HUNK1; HUNKI; MCAP; Protein HUNK1
Gene Symbols BRD4
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23476, 362844
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.