Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BRD7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541544
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541544 Supplier Invitrogen™ Supplier No. PA541544
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KELAQQVTPG DIVSTYGVRK AMGISIPSPV MENNFVDLTE DTEEPKKTDV Sequence homology: Dog: 86%; Guinea Pig: 93%; Horse: 79%; Human: 100%; Rabbit: 79%; Rat: 86%.

BRD7 acts both as coactivator and as corepressor. For example, it activates the Wnt signaling pathway in a DVL1-dependent manner by negatively regulating the GSK3B phosphotransferase activity. In addition, it is required for TP53-mediated cell-cycle arrest in response to oncogene activation. BRD7 promotes acetylation of TP53 at Lys-382, and thereby promotes efficient recruitment of TP53 to target promoters, inhibiting cell cycle progression from G1 to S phase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRD7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BRD7
Gene Accession No. Q9NPI1
Gene Alias 75 kDa bromodomain protein; BP75; BRD7; bromodomain containing 7; bromodomain protein 75 kDa; bromodomain-containing 7; bromodomain-containing 7-like protein; bromodomain-containing protein 7; CELTIX1; NAG4; protein CELTIX-1; protein tyrosine phosphatase, non-receptor type 13 interacting protein; Ptpn13ip
Gene Symbols BRD7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human BRD7 (aa 594-643).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29117
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.