Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRD8 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079780FR
Description
BRD8 Polyclonal antibody specifically detects BRD8 in Human samples. It is validated for Western BlotSpecifications
BRD8 | |
Polyclonal | |
Western Blot | |
bromodomain containing 8, bromodomain-containing protein 8, Skeletal muscle abundant protein, SMAP2, SMAPp120Skeletal muscle abundant protein 2, Thyroid hormone receptor coactivating protein of 120 kDa, trCP120 | |
The immunogen recognized by this antibody maps to a region between residues 25 and 75 of human Bromodomain containing 8 using the numbering given in TrEMBL entry Q969M9 (GeneID 10902). | |
0.1 mL | |
Cellular Markers | |
10902 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction