Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRD8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA581974
Description
Immunogen sequence: EASPESMLSP SHGSNPIEDP LEAETQHKFE MSDSLKEESG TIFGSQIKDA PGEDEEEDGV SEAASLEEPK EEDQGEGYLS EMDNEPPVSE SDDGFSIHNA TLQSHTLADS IPSSPASSQF SVCSEDQEAI QAQKIWKKAI MLVWRAA.
Transcriptional repressor that contribute to growth arrest during terminal macrophage differentiation by repressing target genes involved in Ras-dependent proliferation. Represses MMP1 promoter activity.
Specifications
BRD8 | |
Polyclonal | |
Unconjugated | |
BRD8 | |
2610007E11Rik; 4432404P07Rik; BRD8; bromodomain containing 8; bromodomain-containing protein 8; LOW QUALITY PROTEIN: bromodomain-containing protein 8; p120; Skeletal muscle abundant protein; Skeletal muscle abundant protein 2; SMAP; SMAP2; Thyroid hormone receptor coactivating protein of 120 kDa; trCP120 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10902 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H0E9 | |
BRD8 | |
Recombinant protein corresponding to Human BRD8. Recombinant protein control fragment (Product #RP-95152). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction