Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Active Motif BRD8 / SMAP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Specifications
Antigen | BRD8 / SMAP2 |
---|---|
Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
Applications | ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
IEC-61008
|
Active Motif
61008 |
10 μL | N/A | N/A | N/A | ||||
Description
BRD8 / SMAP2 Polyclonal Antibody for Western Blot, ChIP, ChIP-seq
Transcriptional repressor that contribute to growth arrest during terminal macrophage differentiation by repressing target genes involved in Ras-dependent proliferation. Represses MMP1 promoter activity.Specifications
BRD8 / SMAP2 | |
ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
70mM tris with 0.07mM EDTA, 30% glycerol, 31mM glycine, 105mM NaCl and 0.035% sodium azide; pH 8 | |
Q9H0E9 | |
10902 | |
This BRD8 / SMAP2 antibody was raised against a peptide corresponding to amino acids within the N-terminal region of human BRD8. | |
Antibody |
-20°C, Avoid Freeze/Thaw Cycles | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
BRD8 | |
2610007E11Rik; 4432404P07Rik; BRD8; bromodomain containing 8; bromodomain-containing protein 8; LOW QUALITY PROTEIN: bromodomain-containing protein 8; p120; Skeletal muscle abundant protein; Skeletal muscle abundant protein 2; SMAP; SMAP2; Thyroid hormone receptor coactivating protein of 120 kDa; trCP120 | |
BRD8 | |
Primary | |
Antigen Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title