Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRD9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110872
Description
Immunogen sequence: MGYLKRNGDG SLLYSVVNTA EPDADEEETH PVDLSSLSSK LLPGFTTLGF KDERRNKVTF LSSATTALSM QNNSVFGDLK SDEMELLYS.
Bromodomain containing protein 9 (BRD9) is an uncharacterized protein that contains one bromodomain.
Specifications
| BRD9 | |
| Polyclonal | |
| Unconjugated | |
| BRD9 | |
| AL022779; Brd9; bromodomain containing 9; bromodomain-containing protein 9; DKFZp434D0711; DKFZp686L0539; FLJ43530; LAVS3040; PRO9856; rhabdomyosarcoma antigen MU-RMS-40.8; sarcoma antigen NY-SAR-29; UNQ3040/PRO9856 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 65980 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H8M2 | |
| BRD9 | |
| Recombinant Protein corresonding to Human BRD9. Recombinant protein control fragment (Product #RP-93569). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction