Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRD9 Recombinant Rabbit Monoclonal Antibody (8C4G2)
BRD9 Recombinant Rabbit Monoclonal Antibody (8C4G2)
Supplier: Thermo Scientific MA551462
Description
BRD9 Recombinant Monoclonal Antibody for Western Blot

Specifications
BRD9 | |
Recombinant Monoclonal | |
0.07 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q3UQU0, Q9H8M2 | |
BRD9 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 274-597 of human BRD9 (NP_076413.3). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
Liquid |
ELISA, Western Blot | |
8C4G2 | |
Unconjugated | |
BRD9 | |
AL022779; Brd9; bromodomain containing 9; bromodomain-containing protein 9; DKFZp434D0711; DKFZp686L0539; FLJ43530; LAVS3040; PRO9856; rhabdomyosarcoma antigen MU-RMS-40.8; sarcoma antigen NY-SAR-29; UNQ3040/PRO9856 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
105246, 308067, 65980 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction