Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRDT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541426
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541426 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541426 Supplier Thermo Scientific Supplier No. PA541426
Add to Cart
Add to Cart

BRDT Polyclonal Antibody

BRDT Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen BRDT
Applications Immunohistochemistry (Paraffin), Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene BRDT
Gene Accession No. Q58F21
Gene Alias 7420412D09Rik; BRD6; Brdt; bromodomain testis associated; bromodomain testis-specific protein; bromodomain, testis-specific; bromodomain-containing female sterile homeotic-like protein; Cancer/testis antigen 9; CT9; female sterile homeotic-related gene 3; fi25h02; Fsrg3; LOW QUALITY PROTEIN: bromodomain testis-specific protein; RING3-like protein; si:dkeyp-85h7.3; testis-specific bromodomain protein; wu:fi25h02; wu:fi25h02 protein
Gene Symbols BRDT
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human BRDT (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 676
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.