Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BRDT Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117277
Description
Centrifuge briefly prior to opening.
BRDT is similar to the RING3 protein family. It possesses 2 bromodomain motifs and a PEST sequence , characteristic of proteins that undergo rapid intracellular degradation. The bromodomain is found in proteins that regulate transcription. Two transcript variants encoding the same protein have been found for this gene.
Specifications
BRDT | |
Polyclonal | |
Unconjugated | |
BRDT | |
7420412D09Rik; BRD6; Brdt; bromodomain testis associated; bromodomain testis-specific protein; bromodomain, testis-specific; bromodomain-containing female sterile homeotic-like protein; Cancer/testis antigen 9; CT9; female sterile homeotic-related gene 3; fi25h02; Fsrg3; LOW QUALITY PROTEIN: bromodomain testis-specific protein; RING3-like protein; si:dkeyp-85h7.3; testis-specific bromodomain protein; wu:fi25h02; wu:fi25h02 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114642, 676 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q58F21, Q91Y44 | |
BRDT | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human BRDT. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction