Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrdU Monoclonal Antibody (Bu20a), Active Motif™
SDP

Catalog No. 501993292
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3292 10 μg
50-199-3291 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3292 Supplier Active Motif Supplier No. 61274
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bromodeoxyuridine (5-bromo-2-deoxyuridine, BrdU) is a synthetic nucleoside that is an analogue of thymidine. BrdU is commonly used in the detection of proliferating cells in living tissues, and can be incorporated into the newly synthesized DNA of replicating cells (during the S phase of the cell cycle), substituting for thymidine during DNA replication. Antibodies specific for BrdU can then be used to detect the incorporated chemical, thus indicating cells that were actively replicating their DNA. Binding of the antibody requires denaturation of the DNA, usually by exposing the cells to acid or heat.

Specifications

Antigen BrdU
Applications Immunohistochemistry
Classification Monoclonal
Clone Bu20a
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene FRA10B
Gene Alias 5-Bromo-2-deoxyuridine; Bromodeoxyuridine
Gene Symbols FRA10B
Host Species Mouse
Immunogen This BrdU antibody was raised against Bromodeoxyuridine conjugated to BSA and recognizes BrdU.
Purification Method Protein A
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2432
Target Species Human, Multi-species, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.