Learn More
BrdU Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™

Description
Specifications
Specifications
| Antigen | BrdU |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 140mM HEPES, 32mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 |
| Gene | FRA10B |
| Gene Alias | 5-Bromo-2-deoxyuridine; Bromodeoxyuridine |
| Gene Symbols | FRA10B |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.