Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ BrdU Staining Buffer Set for Flow Cytometry

Catalog No. 501128822
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-8822 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide and formaldehyde
Catalog No. 50-112-8822 Supplier Invitrogen™ Supplier No. 00552500
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Contains Sodium azide and formaldehyde

Contains the buffers needed for optimal BrdU staining with the BU20A monoclonal antibody for flow cytometric analysis. This set, in combination with BrdU and DNase I steps, allows for detection of BrdU pulse-labeled cells, thereby enabling the examination of proliferating cells.

  • Cycling cells are incubated with 5-bromo-2′-deoxyuridine (BrdU), a synthetic analog of thymidine which incorporates into newly synthesized genomic DNA during the S-phase of mitosis
  • Following DNA denaturation, the cells are stained for BrdU incorporation along with any other cell surface and/or intracellular targets of interest
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) BrdU Staining
For Use With (Equipment) Flow Cytometer
Product Type Staining Buffer Set
Product Line eBioscience
Quantity 1 kit

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.