Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ BrdU Staining Kit for Flow Cytometry APC

Catalog No. 501125095
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-5095 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide and formaldehyde
Catalog No. 50-112-5095 Supplier Invitrogen™ Supplier No. 8817660042
Add to Cart
Add to Cart
Contains Sodium azide and formaldehyde

Includes

  • BrdU
  • DNase I
  • Anti-BrdU APC Antibody (clone BU20A)
  • BrdU Staining Buffer Concentrate (4X)
  • Fixation/Permeabilization Diluent

Contains the necessary reagents and buffers for identifying and examining proliferating cells of mammalian species by flow cytometric analysis. This kit is optimized to achieve brighter staining compared to traditional protocols.

  • Cycling cells are incubated with 5-bromo-2′-deoxyuridine (BrdU), a synthetic analog of thymidine which incorporates into newly synthesized genomic DNA during the S-phase of mitosis
  • Following DNA denaturation, the cells are stained for BrdU incorporation along with any other cell surface and/or intracellular targets of interest
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Fluorescence
For Use With (Application) BrdU Staining
For Use With (Equipment) Flow Cytometer
Product Type Staining Kit
Format Kit
Product Line eBioscience
Quantity 100 tests
Can I also stain for intracellular targets such as transcription factors and cytokines using the eBioscience BrdU staining kit?

Yes, primary antibodies against additional intracellular targets can be added alongside the Anti-BrdU antibody. However, it is critical that you empirically determine whether these additional antibodies are compatible with this staining protocol.

Is the BrdU in the eBioscience BrdU staining kit supplied as a sterile formulation for in vivo use?

Yes, the BrdU is supplied as a sterile liquid. The vial should be opened under sterile conditions if it will be used in vivo.

Can I keep the diluted (1 mM) BrdU in the eBioscience BrdU staining kit at 4 degrees C? Can I refreeze it after use?

The stock (32.5 mM) BrdU can be frozen and thawed up to three times. Furthermore, the stock BrdU can be kept at 4 degrees C for up to one week. However, the diluted (1 mM) BrdU should be used immediately.

Have the BrdU Staining kits been tested on proliferating cells of different species?

We have used this kit to detect proliferating human and mouse cells. Other species have yet to be tested but feline, non-human primate and canine should also work.

What are the stopping points during flow cytometric analysis when using a BrdU staining kit?

Samples can be stored at 4 degrees C for up to 16 hours (i.e., overnight) after fixation with BrdU Staining Buffer (with or without washing in Flow Cytometry Staining Buffer) with no adverse effect on performance.

Can I refreeze the DNase I in BrdU Staining kits?

Yes, as supplied, the DNase I can be frozen and thawed up to three times.

How long can I keep the diluted DNase I in BrdU Staining kits at 4 degrees C?

Once diluted to a working concentration, the DNase I can be kept at 4 degrees C for up to 1 week. Longer storage under these conditions will result in a reduction in positive staining.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.