Learn More
Invitrogen™ eBioscience™ BrdU Staining Kit for Flow Cytometry eFluor™ 450
Description
Includes
- BrdU
- DNase I
- Anti-BrdU eFluor 450 Antibody (clone BU20A)
- BrdU Staining Buffer Concentrate (4X)
- Fixation/Permeabilization Diluent
- Cycling cells are incubated with 5-bromo-2’-deoxyuridine (BrdU), a synthetic analog of thymidine which incorporates into newly synthesized genomic DNA during the S-phase of mitosis
- Following DNA denaturation, the cells are stained for BrdU incorporation along with any other cell surface and/or intracellular targets of interest
Specifications
Specifications
| Detection Method | Fluorescence |
| For Use With (Application) | BrdU Staining |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Staining Kit |
| Format | Kit |
| Product Line | eBioscience |
| Quantity | 100 Tests |
Frequently Asked Questions (FAQs)
Yes, primary antibodies against additional intracellular targets can be added alongside the Anti-BrdU antibody. However, it is critical that you empirically determine whether these additional antibodies are compatible with this staining protocol.
Yes, the BrdU is supplied as a sterile liquid. The vial should be opened under sterile conditions if it will be used in vivo.
The stock (32.5 mM) BrdU can be frozen and thawed up to three times. Furthermore, the stock BrdU can be kept at 4 degrees C for up to one week. However, the diluted (1 mM) BrdU should be used immediately.
We have used this kit to detect proliferating human and mouse cells. Other species have yet to be tested but feline, non-human primate and canine should also work.
Samples can be stored at 4 degrees C for up to 16 hours (i.e., overnight) after fixation with BrdU Staining Buffer (with or without washing in Flow Cytometry Staining Buffer) with no adverse effect on performance.
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.