Learn More
Invitrogen™ BRE Polyclonal Antibody

Description
Recommended positive controls: HeLa, 293T, A431, C8D30, rat testis. Predicted reactivity: Mouse (99%), Rat (98%), Zebrafish (85%), Xenopus laevis (91%), Chicken (97%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | BRE |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | BABAM2 |
| Gene Accession No. | Q6P7Q1, Q8K3W0, Q9NXR7 |
| Gene Alias | 6030405P19Rik; AI429776; B830038C02Rik; BABAM2; brain and reproductive organ-expressed (TNFRSF1A modulator); brain and reproductive organ-expressed protein; BRCA1/BRCA2-containing complex subunit 45; BRCA1/BRCA2-containing complex, subunit 4; BRCA1-A complex subunit BRE; BRCC4; BRCC45; BRE; bre {ECO:0000312; BRISC and BRCA1-A complex member 2; RGD:735111} |
| Gene Symbols | BABAM2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.