Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Breast cancer suppressor candidate 1 Antibody (OTI3D6), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271158MFV500
Description
Breast cancer suppressor candidate 1 Monoclonal antibody specifically detects Breast cancer suppressor candidate 1 in Human samples. It is validated for Flow CytometrySpecifications
Breast cancer suppressor candidate 1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
4013 | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
OTI3D6 | |
Flow Cytometry | |
BCSC-1Loss of heterozygosity 11 chromosomal region 2 gene A protein, BCSC1ortholog of mouse AW551984, Breast cancer suppressor candidate 1, LOH11CR2Avon Willebrand factor A domain-containing protein 5A, loss of heterozygosity, 11, chromosomal region 2, gene A, von Willebrand factor A domain containing 5A | |
Full length human recombinant protein of human VWA5A (NP_055437) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction