Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Breast Matched Tumor and Normal Tissue Lysate
SDP

Catalog No. NBP247115 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247115 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP247115 Supplier Novus Biologicals™ Supplier No. NBP247115

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Two matched 1mg/mL vials (100μg/vial) of human breast tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.

The Breast Matched Tumor & Normal Tissue Lysate is derived from human.

Specifications

Tissue Breast
Quantity 2 Vials
Host Species Human
Content And Storage Store at −80°C. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.