Learn More
Eisco™ Breccia Specimen
SCI_ED
Catalog No. S24718 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24718 1 Ea. 2 to 3 cm
S26628 10/Pk. 2 to 3 cm
S24719 12/Pk. 2 to 3 cm
S26606 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24718 Supplier Eisco™ Supplier No. ESNG0029
Add to Cart
Add to Cart
Breccia Specimen has large angular clasts, making it possible to confidently ascertain depositional history of the rock.
  • Large clasts within cement matrix
  • Poorly sorted and rounded clasts indicate deposition in river near clast source
  • For use in learning sedimentary rock classification and identification
  • Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Product Type Sedimentary Rock
For Use With (Application) Lessons in sedimentary rock classification and identification
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Breccia