Description
- Large clasts within cement matrix
- Poorly sorted and rounded clasts indicate deposition in river near clast source
- For use in learning sedimentary rock classification and identification
- Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Specifications
| Product Type | Sedimentary Rock |
| For Use With (Application) | Lessons in sedimentary rock classification and identification |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 12/Pk. |
| Specimen Size | 2 to 3 cm |
| Specimen Type | Breccia |