Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Brequinar sodium
GREENER_CHOICESDP

Catalog No. 619650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
61-965-0 50 mg
61-961-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 61-965-0 Supplier Tocris Bioscience™ Supplier No. 6196/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective DHODH inhibitor.

Brequinar sodium is a potent and selective dihydroorotate dehydrogenase (DHODH) inhibitor (IC50 = ∽20 nM). Causes depletion of pyrimidine nucleotides. Exhibits minimal inhibition against a panel of >400 kinases. Triggers differentiation of AML cell lines (ED50 = ∽ 1 μM. Note uridine concentration effects ED50 value). Causes differentiation and depletion of leukemia-initiating cells in vivo. Antimetabolite.

Specifications

Chemical Name or Material 6-Fluoro-2-(2′-fluoro[1,1′-biphenyl]-4-yl)-3-methyl-4-quinolinecarboxylic acid sodium
CAS 96201-88-6
Assay Percent Range >99%
Quantity 50 mg
Target DHODH Inhibitors
Molecular Formula C23H14F2NO2Na
Formula Weight Observed MW: 397.35
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.