Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brevican Antibody (450616) [mFluor Violet 610 SE], Novus Biologicals™
SDP

Catalog No. NB653749 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB653749 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB653749 Supplier Novus Biologicals Supplier No. FAB40091MFV610
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Brevican Monoclonal antibody specifically detects Brevican in Human samples. It is validated for Immunocytochemistry

Specifications

Antigen Brevican
Applications Immunocytochemistry
Classification Monoclonal
Clone 450616
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias BEHABbrevican core protein, Brain-enriched hyaluronan-binding protein, brevican, Chondroitin sulfate proteoglycan 7, chondroitin sulfate proteoglycan BEHAB, chondroitin sulfate proteoglycan BEHAB/brevican, CSPG7brevican proteoglycan, MGC13038
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Brevican isoform 1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Cellular Signaling, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 63827
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.