Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brevican Antibody (S294A-6), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222401UV
Description
Brevican Monoclonal specifically detects Brevican in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Brevican | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BCAN | |
Fusion protein amino acids 219-655 of rat Brevican | |
0.1 mL | |
63827 | |
Mouse, Rat | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
S294A-6 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
BEHABbrevican core protein, Brain-enriched hyaluronan-binding protein, brevican, Chondroitin sulfate proteoglycan 7, chondroitin sulfate proteoglycan BEHAB, chondroitin sulfate proteoglycan BEHAB/brevican, CSPG7brevican proteoglycan, MGC13038 | |
Mouse | |
Protein G purified | |
Primary | |
Detects approx 140kDa (and smaller due to proteolytic cleavage). | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction