Learn More
Brew Plate™ Blk BiPlate Aerobic
Click to view available options
:
Aerobic Bi-Plate
Description
- Each quadrant made from different media
- Quadrant 1 contains Lee multi-differential agar and Quadrant 2 contains MacConkey agar
Specifications
Specifications
| Media Type | Lee’s Multi-Differential Agar, MacConkey Agar |
| Product Type | Aerobic Bi-Plate |
| Packaging Type | Bulk sleeve |