Learn More
Brew Plate™ Blk Anaerobic BiPlate
Click to view available options
:
Anaerobic Bi-Plate
Description
- Each quadrant made from different media
- Quadrant 1 contains Lee multi-differential agar and Quadrant 2 HSU Lactobacillus/Pediococcus agar
Specifications
Specifications
| Media Type | Lee’s Multi-Differential Agar, Hsu’s Lactobacillus/Pediococcus Agar |
| Product Type | Anaerobic Plate |
| Packaging Type | Bulk sleeve |